Learn More
Sino Biological Recombinant Anti-CD59 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 80299R001F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD59 (rR CD59; Catalog#80299-R08H; P27274; Met1-Asn100) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8261