Learn More
Sino Biological Recombinant Anti-CD59 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80299R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD59 (rR CD59; Catalog#80299-R08H; P27274; Met1-Asn100).
Catalog No. 50-240-8685