Learn More
Sino Biological Recombinant Anti-CD59 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 80299R001P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD59 (rR CD59; Catalog#80299-R08H; P27274; Met1-Asn100) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-9602