Learn More
Sino Biological Rat CD59 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785806
Encompass_Preferred
The Rat CD59 ELISA Pair Set is for the quantitative determination of Rat CD59.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat CD59 coated on a 96-well plate. Standards and samples are added to the wells, and any Rat CD59 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Rat CD59 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat CD59 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85806 Supplier Sino Biological Supplier No. SEK802995
The Rat CD59 ELISA Pair Set is for the quantitative determination of Rat CD59.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat CD59 coated on a 96-well plate. Standards and samples are added to the wells, and any Rat CD59 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Rat CD59 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat CD59 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.