Learn More
Sino Biological Anti-CD59 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80299RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD59 / CD59A / MAC / IP (rR CD59 / CD59A / MAC / IP; Catalog#80299-R08H; P27274; Met1-Asn100). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1901