Learn More
Sino Biological Rat CD63 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80304R04H100
Encompass_Preferred
A DNA sequence encoding the rat CD63 (P28648) (Ala103-Val203) was with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-237-1926