Learn More
Sino Biological Anti-CD68 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80307RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD68 / Macrosialin (rR CD68 / Macrosialin; Catalog#80307-R08H; Q4FZY1; Met1-Ser295). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2789