Learn More
Sino Biological Anti-CD68 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80307T2420
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD68 (Catalog#80307-R08H; Q4FZY1; Met1-Ser295). CD68 specific IgG was purified by Rat CD68 affinity chromatography.
Catalog No. 50-255-2787