Learn More
Sino Biological Recombinant Rat CD68 Protein (hFc Tag)
Supplier:  Sino Biological 80307R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD68 (Q4FZY1) (Met1-Ser295) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3761