Learn More
Sino Biological Anti-CD79B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80326T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD79B (Catalog#80326-R08H; O70153; Met1-Asp158). CD79B specific IgG was purified by Rat CD79B affinity chromatography.
Catalog No. 50-255-2783