Learn More
Sino Biological Rat CD79B Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80326R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD79B (O70153) (Met1-Asp158) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1619