Learn More
Sino Biological Recombinant Rat B7-1/CD80 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 80027R08H1
Encompass_Preferred
A DNA sequence encoding the Rat CD80 (AAB60503.1) (Met1-Gln248) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-241-1612