Learn More
Sino Biological Rat CD83 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80333R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD83 (B2GV95) (Met1-Ala134) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2765