Learn More
Sino Biological Recombinant Anti-C1qR1/CD93 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 80207R138F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD93/C1qR (rR CD93/C1qR; Catalog#80207-R08H; Q9ET61; Met1-Lys571) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8643