Learn More
Sino Biological Anti-C1qR1/CD93 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80207RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD93 / C1qR (Catalog#80207-R08H; Q9ET61; Met1-Lys571). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2211