Learn More
Sino Biological Anti-C1qR1/CD93 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80207T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CD93/C1qR (rR CD93/C1qR; Catalog#80207-R08H; Q9ET61; Met1-Lys571). CD93/C1qR specific IgG was purified by Rat CD93/C1qR affinity chromatography.
Catalog No. 50-240-8354