Learn More
Sino Biological Anti-E-Cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80274RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat E-Cadherin / CDH1 / E-cad / CD324 (rR E-Cadherin / CDH1 / E-cad / CD324; Catalog#80274-R08H; Q9R0T4; Met1-Ala713). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1052