Learn More
Sino Biological Rat CDH1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 80274T50100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat E-Cadherin / CDH1 / E-cad / CD324 (rR E-Cadherin / CDH1 / E-cad / CD324; Catalog#80274-R08H; Q9R0T4; Met1-Ala713). E-Cadherin / CDH1 / E-cad / CD324 specific IgG was purified by Rat E-Cadherin / CDH1 / E-cad / CD324 affinity chromatography.
Catalog No. 50-237-3131