Learn More
Sino Biological Rat CDH1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80274R08H100
Encompass_Preferred
A DNA sequence encoding the rat CDH1 (Q9R0T4) (Met1-Ala713) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2962