Learn More
Sino Biological Anti-VE-Cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80276T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat VE-Cadherin / CD144 (Catalog#80276-R08H; NP_001100877.1; Met1-Gln585). VE-Cadherin / CD144 specific IgG was purified by Rat VE-Cadherin / CD144 affinity chromatography.
Catalog No. 50-240-8456