Learn More
Sino Biological Anti-Cadherin-6/K-cadherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80277T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat K Cadherin / CDH6 (rR K Cadherin / CDH6; Catalog#80277-R08H; EDL82944.1; Met1-Ala615). K Cadherin / CDH6 specific IgG was purified by Rat K Cadherin / CDH6 affinity chromatography.
Catalog No. 50-240-9581