Learn More
Sino Biological Rat CDH6 Protein 200ug (Sino Biological)
Supplier:  Sino Biological 80277R08H200
Encompass_Preferred
A DNA sequence encoding the rat CDH6 (EDL82944.1) (Met1-Ala615) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3841