Learn More
Sino Biological Anti-Cadherin 8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80278T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Cadherin-8 / CDH8 (rR Cadherin-8 / CDH8; Catalog#80278-RCCH; NP_445845.2; Met1-Met621). Cadherin-8 / CDH8 specific IgG was purified by Rat Cadherin-8 / CDH8 affinity chromatography.
Catalog No. 50-255-2790