Learn More
Sino Biological Rat CDH8 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 80278RCCH50
Encompass_Preferred
A DNA sequence encoding the rat CDH8 (NP_445845.2)(Met1-Met621) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-237-3532