Learn More
Sino Biological Anti-CEACAM-1/CD66a Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 80224MM03P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CEACAM‑1/CD66a (rR CEACAM‑1/CD66a; Catalog#80224-R08H; Met1-Ser422) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-9068