Learn More
Lonza Walkersville Rat Cerebellar Granular Neuron
Catalog No. 5000496422
Encompass
Clonetics Rat Cerebellar Granular Cells are isolated from cerebellar cortex of adult Sprague Dawley rats at postnatal day 8 Cerebellar granular cells utilize glutamate as their neurotransmitter and as such are the only excitatory neurons within the cerebellar cortex All other neuronal cell types employ GABA as a neurotransmitter Granular cells arise during the latter stages of neuronal development of the cerebellar cortex and are the most abundant single neuronal type in the mammalian brain These are highly homogeneous populations of granular cells containing less than 5% GABAergic neurons as assessed by GABA immunoreactivity Recommended Media PNGM-A BulletKit (CC-4512)
Catalog No. 50-004-96422 Supplier Lonza Walkersville Supplier No. RCB503
Clonetics Rat Cerebellar Granular Cells are isolated from cerebellar cortex of adult Sprague Dawley rats at postnatal day 8 Cerebellar granular cells utilize glutamate as their neurotransmitter and as such are the only excitatory neurons within the cerebellar cortex All other neuronal cell types employ GABA as a neurotransmitter Granular cells arise during the latter stages of neuronal development of the cerebellar cortex and are the most abundant single neuronal type in the mammalian brain These are highly homogeneous populations of granular cells containing less than 5% GABAergic neurons as assessed by GABA immunoreactivity Recommended Media PNGM-A BulletKit (CC-4512)