Learn More
Biomatik Corporation Rat Ceruloplasmin (CP) CLIA Kit
Supplier:  Biomatik Corporation EKU09790
Encompass_Preferred
Rat Ceruloplasmin (CP) CLIA Kit for detecting CP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.55-400ng/ml. Sensitivity: 0.25ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P13635. Target Synonyms: CP2; CER; Caeruloplasmin; Ferroxidase; Oxygen Oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5614