Learn More
Medchemexpress LLC HY-P0209 1mg Medchemexpress, Rat CGRP-(8-37) CAS:129121-73-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02091MG
Encompass_Preferred
Medchemexpress, HY-P0209 1mg Rat CGRP-(8-37) CAS:129121-73-9 Rat CGRP-(8-37) (VTHRLAGLLSRSGGVVKDNFVPTNVGSEAF) is a highly selective CGRP receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4051