Learn More
Raybiotech Inc C-Series Rat Chemokine Array 1 Kit.
Supplier: Raybiotech Inc AARCHE14
Detects 15 rat Chemokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: MIP-3 alpha, CINC-1, CINC-2, CINC-3, CTACK, Eotaxin-1, Fractalkine, IL-1 alpha, LIX, MCP-1, MIG, MIP-1 beta, MIP-1 alpha, TCK-1, TNF alpha