Learn More
Biomatik Corporation Rat Chemokine (C-X-C motif) ligand 7 (CXCL7) CLIA Kit
Supplier:  Biomatik Corporation EKU09788
Encompass_Preferred
Rat Chemokine (C-X-C motif) ligand 7 (CXCL7) CLIA Kit for detecting C-X-C motif in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 4.12-3,000pg/ml. Sensitivity: 1.75pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q99ME0. Gene ID: 246358. Target Synonyms: CXCL7; PPBP; PBP; B-TG1; CTAP-III; CTAP3; CTAPIII; LA-PF4; LDGF; MDGF; NAP2; SCYB7; TC1; TC2; TGB1; THBGB1; Pro-Platelet Basic Protein; Beta-Thromboglobulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8434