Learn More
BIOHIPPO Rat CHGA Microsample
SDP
Catalog No. 5000313761
Encompass
Rat CHGA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CHGA in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 34 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CHGA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CHGA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CHGA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13761 Supplier BIOHIPPO Supplier No. BHE15211232
Rat CHGA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CHGA in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 34 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CHGA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CHGA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CHGA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color