Learn More
Biomatik Corporation Rat Choline Acetyltransferase (ChAT) CLIA Kit
Supplier:  Biomatik Corporation EKU10023
Encompass_Preferred
Rat Choline Acetyltransferase (ChAT) CLIA Kit for detecting ChAT in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P32738. Target Synonyms: CMS1A; CMS1A2; CHOACTase; Choline acetylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6833