Learn More
Biomatik Corporation Rat Ciliary Neurotrophic Factor (CNTF) CLIA Kit
Supplier:  Biomatik Corporation EKU09430
Encompass_Preferred
Rat Ciliary Neurotrophic Factor (CNTF) CLIA Kit for detecting CNTF in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.61pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P20294. Gene ID: 25707. Target Synonyms: HCNTF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6561