Learn More
Raybiotech Inc Rat CINC-1 (CXCL1) ELISA Kit
Supplier:  Raybiotech Inc ELRCINC11
Encompass_Preferred
For cell culture supernatants, Heparin and/or EDTA treated plasma, and serum samples. Citrate is not recommended. Sensitivity: 300 pg/ml in Assay Diluent D and 50 pg/ml in Assay Diluent B. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with any of the cytokines tested: rat CINC-2, CINC-3, CNTF, IL-1 alpha, IL-1 beta, IL-6, IL-10, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, beta-NGF, TIMP-1, TNF-alpha, VEGF.
Catalog No. 50-195-0224