Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat CINC-2 ELISA Kit

Catalog No. ERA7RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ERA7RBX5 5 x 96 Tests
ERA7RB 96 Tests
50-254-5408 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ERA7RBX5 Supplier Invitrogen™ Supplier No. ERA7RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Rat CINC-2 quantitates rat CINC-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat CINC-2.

CXCL3 may play a role in inflammation and exert its effects on endothelial cells in an autocrine fashion. All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P19876, Q10746
Assay Range 17-4000 pg/mL
Assay Sensitivity 17 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human, Rat
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 3; Cinc2; CINC-2; CINC-2b; CXC; CXC motif chemokine; C-X-C motif chemokine 3; C-X-C motif chemokine ligand 3; CXCL; Cxcl3; cytokine-induced neutrophil chemoattractant 2; cytokine-induced neutrophil chemoattractant-2; Dcip1; Dendritic cell inflammatory protein 1; Gm1960; gro gamma; GRO3; GRO3 oncogene; GROG; GRO-gamma; GRO-gamma(1-73); GRO-gamma(5-73); Growth-regulated protein gamma; macrophage inflammatory protein 2-alpha/beta; Macrophage inflammatory protein 2-beta; melanoma growth stimulatory activity gamma; MGSA gamma; MIP2-alpha/beta; MIP2B; MIP-2b; MIP2-beta; SCYB3C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 3; Cinc2; CINC-2; CINC-2b; CXC; CXC motif chemokine; C-X-C motif chemokine 3; C-X-C motif chemokine ligand 3; CXCL; Cxcl3; cytokine-induced neutrophil chemoattractant 2; cytokine-induced neutrophil chemoattractant-2; Dcip1; Dendritic cell inflammatory protein 1; Gm1960; gro gamma; GRO3; GRO3 oncogene; GROG; GRO-gamma; GRO-gamma(1-73); GRO-gamma(5-73); Growth-regulated protein gamma; macrophage inflammatory protein 2-alpha/beta; Macrophage inflammatory protein 2-beta; melanoma growth stimulatory activity gamma; MGSA gamma; MIP2-alpha/beta; MIP2B; MIP-2b; MIP2-beta; SCYB3;
Gene ID (Entrez) 171551, 2921
Gene Symbol Cxcl3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target CXCL3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.