Learn More
Sino Biological Rat CKMT2 Protein 50ug (Sino Biological)
Catalog No. 502374192
Encompass_Preferred
A DNA sequence encoding the rat Ckmt2 (NP_001121124.1) (Asp40-Lys419) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-4192 Supplier Sino Biological Supplier No. SEK1059415
A DNA sequence encoding the rat Ckmt2 (NP_001121124.1) (Asp40-Lys419) was expressed with a polyhistidine tag at the N-terminus.