Learn More
American Research Products Inc Rat Clara cell protein CC16 E
Catalog No. 5000329721
Encompass
Rat Clara cell protein CC16 ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29721 Supplier American Research Products Inc Supplier No. CSBE08681R
Rat Clara cell protein CC16 ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich