Learn More
Biomatik Corporation Rat Claudin 1 (CLDN1) ELISA Kit
Supplier:  Biomatik Corporation EKU03203
Encompass_Preferred
Rat Claudin 1 (CLDN1) ELISA Kit for detecting CLDN1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56745. Gene ID: 65129. Target Synonyms: CLD1; ILVASC; SEMP1; Senescence-Associated Epithelial Membrane Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9058