Learn More
Biomatik Corporation Rat Claudin 16 (CLDN16) ELISA Kit
Supplier:  Biomatik Corporation EKU03205
Encompass_Preferred
Rat Claudin 16 (CLDN16) ELISA Kit for detecting CLDN16 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.53ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91Y55. Target Synonyms: PCLN1; HOMG3; Paracellin-1; Hypomagnesemia 3,With Hypercalciuria And Nephrocalcinosis. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7566