Learn More
Sino Biological Rat CLEC4B2 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80265R07H100
Encompass_Preferred
A DNA sequence encoding the rat CLEC4B2 (Q67DU9) (Met44-Leu208) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-2905