Learn More
Sino Biological Anti-MDL-1/CLEC5A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80259T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CLEC5A / MDL1 / MDL-1 (Catalog#80259-R07H; F7GAV1; Val30-Lys190). CLEC5A / MDL1 / MDL-1 specific IgG was purified by Rat CLEC5A / MDL1 / MDL-1 affinity chromatography.
Catalog No. 50-255-2792