Learn More
BIOHIPPO Rat CLU Microsample
SDP
Catalog No. 5000315180
Encompass
Rat CLU Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CLU in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 57-100 g/mL with a sensitivity of 0 56 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CLU Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CLU Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CLU biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15180 Supplier BIOHIPPO Supplier No. BHE15212518
Rat CLU Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CLU in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 57-100 g/mL with a sensitivity of 0 56 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CLU Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CLU Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CLU biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color