Learn More
American Research Products Inc Rat Cluster of Differentiation
Catalog No. 5000329839
Encompass
Rat Cluster of Differentiation 68 CD68 ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29839 Supplier American Research Products Inc Supplier No. CSBE13297R
Rat Cluster of Differentiation 68 CD68 ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich