Learn More
American Research Products Inc Rat cluster of differentiation
Catalog No. 5000326388
Encompass
Rat cluster of differentiation 4 CD4 ELISA Kit from CUSABIO Detection Range 0 78 U/mL-50 U/mL Sensitivity 0 195 U/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26388 Supplier American Research Products Inc Supplier No. CSBE12928R
Rat cluster of differentiation 4 CD4 ELISA Kit from CUSABIO Detection Range 0 78 U/mL-50 U/mL Sensitivity 0 195 U/mL Sample serum plasma tissue homogenates Method Sandwich