Learn More
Biomatik Corporation Rat Cluster Of Differentiation 36 (CD36) ELISA Kit
Supplier:  Biomatik Corporation EKU06709
Encompass_Preferred
Rat Cluster Of Differentiation 36 (CD36) ELISA Kit for detecting CD36 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07969. Target Synonyms: GP4; FAT; GP3B; GPIV; PASIV; SCARB3; GP88; GpIIIb; Glycoprotein IIIb; Collagen Type I Receptor; Thrombospondin Receptor; Fatty acid translocase; Platelet collagen receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6966