Learn More
Biomatik Corporation Rat Cluster Of Differentiation 30 Ligand (CD30L) ELISA Kit
Supplier:  Biomatik Corporation EKU09408
Encompass_Preferred
Rat Cluster Of Differentiation 30 Ligand (CD30L) ELISA Kit for detecting CD30L in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 125-8000pg/ml. Sensitivity: 50pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: M0R3V3. Gene ID: 683163. Target Synonyms: CD153; TNFSF8; CD30-L; Tumor Necrosis Factor Ligand Superfamily Member 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6990