Learn More
Biomatik Corporation Rat Cluster of Differentiation 90 (CD90) ELISA Kit
Supplier:  Biomatik Corporation EKU07654
Encompass_Preferred
Rat Cluster of Differentiation 90 (CD90) ELISA Kit for detecting CD90 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01830. Gene ID: 24832. Target Synonyms: Thy1; CDw90; THY mocyte differentiation antigen 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8738