Learn More
Immunology Consultants Rat Clusterin ELISA Kit
SDP
Supplier:  Immunology Consultants E25CLS
Encompass_Preferred
Clusterin plays key roles in protein homeostasis/proteostasis, and the modulation of pro-survival signaling networks. The Rat Clusterin (CLS) test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring CLS in ratbiological samples.
Catalog No. 50-240-6522