Learn More
Meso Scale Discovery Rat Clusterin Kit
SDP
Catalog No. 5000078341
Encompass
The Meso Scale Discovery Rat Clusterin Kit is an immunoassay that quantitatively measures clusterin in rat serum plasma and other biological samples This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and rapid detection of clusterin The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrator and sample diluent The assay has a typical sensitivity of 1-2 ng/mL and a broad dynamic range up to 10 000 ng/mL This product enables researchers to accurately quantify clusterin levels in rat models
Catalog No. 50-000-78341 Supplier Meso Scale Discovery Supplier No. K153MGC1
The Meso Scale Discovery Rat Clusterin Kit is an immunoassay that quantitatively measures clusterin in rat serum plasma and other biological samples This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and rapid detection of clusterin The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrator and sample diluent The assay has a typical sensitivity of 1-2 ng/mL and a broad dynamic range up to 10 000 ng/mL This product enables researchers to accurately quantify clusterin levels in rat models