Learn More
Meso Scale Discovery Rat Clusterin Kit
SDP
Catalog No. 5000081397
Encompass
The Meso Scale Discovery Rat Clusterin Kit is an immunoassay for measuring clusterin in rat serum plasma and other samples It uses MSD s electrochemiluminescent detection for high sensitivity wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibody a detection antibody with an electrochemiluminescent label and calibrators for a standard curve It is specific to rat clusterin with no cross-reactivity with other species and can perform up to 40 tests in duplicate in about 3 5 hours
Catalog No. 50-000-81397 Supplier Meso Scale Discovery Supplier No. K153MGC4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Rat Clusterin Kit is an immunoassay for measuring clusterin in rat serum plasma and other samples It uses MSD s electrochemiluminescent detection for high sensitivity wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibody a detection antibody with an electrochemiluminescent label and calibrators for a standard curve It is specific to rat clusterin with no cross-reactivity with other species and can perform up to 40 tests in duplicate in about 3 5 hours